Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Venlafaxine (hydrochloride) | 99300-78-4 | MFCD03658865 | >99.9% | 100 MG
SDP

Catalog No. 5000405790
Encompass_Preferred

Venlafaxine hydrochloride is an analytical standard used in research and analytical applications. It functions as an orally active, potent serotonin (5-HT) and norepinephrine (NE) reuptake dual inhibitor, recognized as an antidepressant. This compound is suitable for qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.

  • Analytical standard for research and analysis
  • Potent serotonin and norepinephrine reuptake inhibitor
  • Antidepressant properties
  • Suitable for HPLC, GC, and MS applications
  • Molecular formula: C₁₇H₂₈ClNO₂
  • Molecular weight: 313.86
  • Appears as a white to off-white solid
  • Recommended storage: 4°C, sealed, away from light and moisture

Catalog No. 50-004-05790 Supplier Medchemexpress LLC Supplier No. HYB0196AR100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venlafaxine hydrochloride is an analytical standard used in research and analytical applications. It functions as an orally active, potent serotonin (5-HT) and norepinephrine (NE) reuptake dual inhibitor, recognized as an antidepressant. This compound is suitable for qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.

  • Analytical standard for research and analysis
  • Potent serotonin and norepinephrine reuptake inhibitor
  • Antidepressant properties
  • Suitable for HPLC, GC, and MS applications
  • Molecular formula: C₁₇H₂₈ClNO₂
  • Molecular weight: 313.86
  • Appears as a white to off-white solid
  • Recommended storage: 4°C, sealed, away from light and moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.