Learn More
Worthington Biochemical Corporation VENOM PHOSPHODIESTERASE I
SDP
Catalog No. NC9535148
Encompass
Purified by the method of Williams, Sung and Laskowski, JBC, 236, 1130 (1961). Further treated to inactivate contaminating 5'-nucleotidase activity according to Sulkowski and Laskowski: BBA, 240, 443 (1961). Lyophilized in vials.
Catalog No. NC9535148 Supplier Worthington Biochemical Corporation Supplier No. LS003926
Purified by the method of Williams, Sung and Laskowski, JBC, 236, 1130 (1961). Further treated to inactivate contaminating 5'-nucleotidase activity according to Sulkowski and Laskowski: BBA, 240, 443 (1961). Lyophilized in vials.