Learn More
MINERVA BIOLABS INC Venor®GeM OneStep, 25 Tests
Catalog No. NC3661720
Encompass_Preferred
Venor®GeM OneStep is a kit for direct detection of mycoplasma contaminations in cell cultures, cell culture media, and other biological matrices by conventional PCR. Designed to require minimal pipetting, the kit includes all reagents required for PCR: primers, nucleotides, polymerase and the internal amplification control, which are provided in a ready-to-use lyophilized reaction mix. Upon rehydration with the included buffer, the mix is ready to be dispensed into the reaction tubes according to the sample number. Only samples or controls need to be added prior to PCR.
Catalog No. NC3661720 Supplier MINERVA BIOLABS INC Supplier No. 118025
Venor®GeM OneStep is a kit for direct detection of mycoplasma contaminations in cell cultures, cell culture media, and other biological matrices by conventional PCR. Designed to require minimal pipetting, the kit includes all reagents required for PCR: primers, nucleotides, polymerase and the internal amplification control, which are provided in a ready-to-use lyophilized reaction mix. Upon rehydration with the included buffer, the mix is ready to be dispensed into the reaction tubes according to the sample number. Only samples or controls need to be added prior to PCR.