Learn More
Air Science Usa Flammable Cabinet Filtration Unit
SDP
Catalog No. NC3957387
Encompass_Preferred
Vent BOX Low-profile, completely portable, no ductwork required.Compact size, easily positioned on shallow safety cabinets.Modular design permits multiple configurations.Universal connections allow retrofit to almost all available flammable and safety cabinets on the market
Catalog No. NC3957387 Supplier Air Science Usa Supplier No. VB60A
Vent BOX Low-profile, completely portable, no ductwork required.Compact size, easily positioned on shallow safety cabinets.Modular design permits multiple configurations.Universal connections allow retrofit to almost all available flammable and safety cabinets on the market