Learn More
Thomson Instrument Company Ultra Yield 1.5 & 2.5L Vent Cap
SDP
Catalog No. NC1277101
Encompass_Preferred
Thomson’s Vented Screw Caps are sterile, hydrophobic 0.2 µm ventilated closures that replace loose caps and foams—delivering high oxygen transfer while maintaining a clean microbial barrier. As part of the Ultra Yield System, teams report markedly higher cell densities and—when paired with Ultra Yield Flasks and Plasmid+ Media—plasmid yields up to 610%.Key features
•Integrated 0.2 µm hydrophobic vent for high gas exchange with reliable contamination control
•Leak-tight screw closure engineered for high-rpm orbital shaking
•Sterile, ready-to-use to reduce prep time and handling risk
•Sized to match Ultra Yield Flasks (125 mL to 2.5 L) for a secure, consistent fit
•Durable construction for consistent performance across culture runs
• Made in the USA
Catalog No. NC1277101 Supplier Thomson Instrument Company Supplier No. 899566
Thomson’s Vented Screw Caps are sterile, hydrophobic 0.2 µm ventilated closures that replace loose caps and foams—delivering high oxygen transfer while maintaining a clean microbial barrier. As part of the Ultra Yield System, teams report markedly higher cell densities and—when paired with Ultra Yield Flasks and Plasmid+ Media—plasmid yields up to 610%.Key features
•Integrated 0.2 µm hydrophobic vent for high gas exchange with reliable contamination control
•Leak-tight screw closure engineered for high-rpm orbital shaking
•Sterile, ready-to-use to reduce prep time and handling risk
•Sized to match Ultra Yield Flasks (125 mL to 2.5 L) for a secure, consistent fit
•Durable construction for consistent performance across culture runs
• Made in the USA