Learn More
Thomson Instrument Company Vent Caps for Optimum Growth 500mL Flask, Sterile
SDP
Catalog No. 502221347
Encompass_Preferred
Designed for use with Optimum Growth 500 mL Flasks, this sterile vent cap features a 0.2 µm PTFE membrane for enhanced aeration while maintaining sterility. Ideal for cell culture and microbial applications requiring gas exchange, it offers reliable performance and secure flask compatibility.Flask Compatibility: 500mLMembrane: PTFEPore Size: 0.2µmSterility (SAL): 10⁻⁶ Made in the USA
Catalog No. 50-222-1347 Supplier Thomson Instrument Company Supplier No. 899112
Designed for use with Optimum Growth 500 mL Flasks, this sterile vent cap features a 0.2 µm PTFE membrane for enhanced aeration while maintaining sterility. Ideal for cell culture and microbial applications requiring gas exchange, it offers reliable performance and secure flask compatibility.Flask Compatibility: 500mLMembrane: PTFEPore Size: 0.2µmSterility (SAL): 10⁻⁶ Made in the USA