Learn More
Thomson Instrument Company Vent Caps for Ultra Yield 125mL Flask, Sterile
SDP
Catalog No. NC1748882
Encompass_Preferred
Thomson’s Vented Screw Caps are sterile, hydrophobic 0.2 µm ventilated closures that replace loose caps and foams—delivering high oxygen transfer while maintaining a clean microbial barrier. As part of the Ultra Yield System, teams report markedly higher cell densities and—when paired with Ultra Yield Flasks and Plasmid+ Media—plasmid yields up to 610%.Key features
•Integrated 0.2 µm hydrophobic vent for high gas exchange with reliable contamination control
•Leak-tight screw closure engineered for high-rpm orbital shaking
•Sterile, ready-to-use to reduce prep time and handling risk
•Sized to match Ultra Yield Flasks (125 mL to 2.5 L) for a secure, consistent fit
•Durable construction for consistent performance across culture runs
• Made in the USA
Catalog No. NC1748882 Supplier Thomson Instrument Company Supplier No. 899109
Thomson’s Vented Screw Caps are sterile, hydrophobic 0.2 µm ventilated closures that replace loose caps and foams—delivering high oxygen transfer while maintaining a clean microbial barrier. As part of the Ultra Yield System, teams report markedly higher cell densities and—when paired with Ultra Yield Flasks and Plasmid+ Media—plasmid yields up to 610%.Key features
•Integrated 0.2 µm hydrophobic vent for high gas exchange with reliable contamination control
•Leak-tight screw closure engineered for high-rpm orbital shaking
•Sterile, ready-to-use to reduce prep time and handling risk
•Sized to match Ultra Yield Flasks (125 mL to 2.5 L) for a secure, consistent fit
•Durable construction for consistent performance across culture runs
• Made in the USA