Learn More
New England Biolabs, Inc. Vent DNA Polymerase – 1000 units
SDP
Catalog No. 50811672
Encompass
Vent DNA Polymerase is a high-fidelity thermophilic DNA polymerase. The fidelity of Vent DNA Polymerase is 5-15-fold higher than that observed for Taq DNA Polymerase. This high fidelity derives in part from an integral 3' to 5' proofreading exonuclease activity in Vent DNA Polymerase. Greater than 90% of the polymerase activity remains following a 1 hour incubation at 95C.
Catalog No. 50-811-672 Supplier New England Biolabs, Inc. Supplier No. M0254L
Vent DNA Polymerase is a high-fidelity thermophilic DNA polymerase. The fidelity of Vent DNA Polymerase is 5-15-fold higher than that observed for Taq DNA Polymerase. This high fidelity derives in part from an integral 3' to 5' proofreading exonuclease activity in Vent DNA Polymerase. Greater than 90% of the polymerase activity remains following a 1 hour incubation at 95C.