Learn More
New England Biolabs, Inc. Vent DNA Polymerase – 200 units
SDP
Catalog No. 50811673
Encompass
Vent DNA Polymerase is a high-fidelity thermophilic DNA polymerase. The fidelity of Vent DNA Polymerase is 5-15-fold higher than that observed for Taq DNA Polymerase. This high fidelity derives in part from an integral 3' to 5' proofreading exonuclease activity in Vent DNA Polymerase. Greater than 90% of the polymerase activity remains following a 1 hour incubation at 95C.
Catalog No. 50-811-673 Supplier New England Biolabs, Inc. Supplier No. M0254S
Vent DNA Polymerase is a high-fidelity thermophilic DNA polymerase. The fidelity of Vent DNA Polymerase is 5-15-fold higher than that observed for Taq DNA Polymerase. This high fidelity derives in part from an integral 3' to 5' proofreading exonuclease activity in Vent DNA Polymerase. Greater than 90% of the polymerase activity remains following a 1 hour incubation at 95C.