Learn More
New England Biolabs, Inc. Vent (exo-) DNA Polymerase – 1000 units
SDP
Catalog No. 50811676
Encompass
Vent (exo-) DNA Polymerase has been genetically engineered to eliminate the 3 5 proofreading exonuclease activity associated with Vent DNA Polymerase. This is the preferred form for high-temperature dideoxy sequencing reactions and for high yield primer extension reactions. The fidelity of polymerization by this form is reduced to a level about 2-fold higher than that of Taq DNA Polymerase.
Catalog No. 50-811-676 Supplier New England Biolabs, Inc. Supplier No. M0257L
Vent (exo-) DNA Polymerase has been genetically engineered to eliminate the 3 5 proofreading exonuclease activity associated with Vent DNA Polymerase. This is the preferred form for high-temperature dideoxy sequencing reactions and for high yield primer extension reactions. The fidelity of polymerization by this form is reduced to a level about 2-fold higher than that of Taq DNA Polymerase.