Learn More
Thomas Scientific Vent Filter, 0.22um, 1/4"' Tubing, Non-Sterile
SDP
Supplier:  Thomas Scientific 1170A22
Encompass
Foxx Life Science Carboy Vent Filters are used for hydrophobic venting and solvent filtration applications. The PTFE filter membrane is ideal for maintaining purified water stored in carboys, or for sterile venting during a slow exhaust/liquid autoclave cycle of EZgrip® carboys. All materials comply with USP Class VI standards, and are autoclavable up to 10 times. Hydrophobic PTFE membrane is reinforced with polypropylene housing and features a stepped hosebarb. Available in Non-sterile 1.00µm, 0.2µm pore sizes with platinum-cured silicone tubing, or in bulk packs (without tubing).
Catalog No. 50-145-8496