Learn More
Bellco Glass, Inc. Vent Filter, 0.22um, Non-Sterile, 5/PK
SDP
Supplier:  Bellco Glass, Inc. 452800582
Encompass_Preferred
EZFlow 0.2µm PTFE Vent Filter, 50mm PP Housing, 1/4" to 1/2" Stepped Barb, Non Sterile, 5/PK Carboy Vent Filters are used for hydrophobic venting and solvent filtration applications. The PTFE filter membrane is ideal for maintaining purified water stored in carboys, or for sterile venting during a slow exhaust/liquid autoclave cycle of EZgrip carboys.Hydrophobic PTFE membrane is reinforced with polypropylene housing and features a stepped hosebarb. Available in Non-sterile 1.00µm, 0.2µm pore sizes with platinum-cured silicone tubing, or in bulk packs (without tubing).SPECIFICATIONSHydrophobic PTFE membrane reinforced in polypropyleneAvailable in 1.00µm and 0.2µm pore sizesAutoclavable up to 10 timesCasing Material: PolypropyleneFilter Material: PTFEFilter Pore Size: 0.2µmMade in USA
Catalog No. 50-255-0180