Learn More
Bellco Glass, Inc. Vent Filter,0.22 μm,62 mm diameter, Individually Wrapped, Sterile, 1/pk, 5/cs
SDP
Supplier:  Bellco Glass, Inc. 451400106
Encompass_Preferred
Vent Filter,0.22 μm,62 mm diameter, Individually Wrapped, Sterile, 1/pk, 5/cs. Using with BioFactory accessories during cell culture process could simplify the liquid transfer process,meanwhile effectively reduce the risk of cell contamination. • Sterilized• Individually Wrapped in Plastic Sterile Bag
Catalog No. 50-232-9270