Learn More
Chemglass Life Sciences Vent Heater, 4 Watt
SDP
Catalog No. 5000083132
Encompass
Filter vent heater used to keep 40x70mm, rectangular vent filters free of harmful condensation that can compromise cultures.
Catalog No. 50-000-83132 Supplier Chemglass Life Sciences Supplier No. CLS1200FH
Filter vent heater used to keep 40x70mm, rectangular vent filters free of harmful condensation that can compromise cultures.