Learn More
Ankom Technology Vent Valve Adapter
SDP
Catalog No. NC1875486
Encompass
Vent Valve Adapter is used to attach the 7043 Syringe to the vent valve tube to aid in cleaning the vent valve on a Gas Production Module.
Catalog No. NC1875486 Supplier Ankom Technology Supplier No. RF22.5
Vent Valve Adapter is used to attach the 7043 Syringe to the vent valve tube to aid in cleaning the vent valve on a Gas Production Module.