Learn More
Waters Corp Vent Valve Cartridge, HPLC, UPLC, Mixer
Catalog No. 502392972 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement vent valve cartridge is utilized on binary solvent managers and sample managers.
  • Used in HPLC and UPLC systems.
  • Functions as a mixer.
  • One unit per package.
  • Replacement is scheduled as required.
  • Accessory for quaternary solvent managers.
  • Accessory for binary solvent managers.
  • Included in performance maintenance kits.
  • Also known as a vent valve/trap valve cartridge.
Catalog No. 50-239-2972 Supplier Waters Corp Supplier No. 700002660
This replacement vent valve cartridge is utilized on binary solvent managers and sample managers.
  • Used in HPLC and UPLC systems.
  • Functions as a mixer.
  • One unit per package.
  • Replacement is scheduled as required.
  • Accessory for quaternary solvent managers.
  • Accessory for binary solvent managers.
  • Included in performance maintenance kits.
  • Also known as a vent valve/trap valve cartridge.