Learn More
Perkin Elmer US LLC Vented Autosampler Aluminum Sample Pans - 30 µL, Qty. 400
Catalog No. 502112428
Encompass_Preferred
These volatile pans and covers are suitable for use with DSC autosamplers, but are also suitable for use with non-robotic systems. Most of these pans and covers can be crimped using the Universal Crimper (B0139005).
Catalog No. 50-211-2428 Supplier Perkin Elmer US LLC Supplier No. B0143018
Add to Cart
Add to Cart
These volatile pans and covers are suitable for use with DSC autosamplers, but are also suitable for use with non-robotic systems. Most of these pans and covers can be crimped using the Universal Crimper (B0139005).