Learn More
Chemglass Life Sciences Vented Cap Label for Reaction Tubes, 5 Labels per Row (1 thru 5), 50 Labels per Sheet
SDP
Catalog No. 501532366
Encompass_Preferred
Vented Cap Label for Reaction Tubes, 5 Labels per Row (1 thru 5), 50 Labels per Sheet. Labels are used to block from 1 to 5 openings on the stopper of the 50mL bio reaction tubes to control the amount of CO2 exchange and evaporation in the tube. Sheets have 8 each of white Mylar labels from numbers 1 to 5, and are 10 sets of labels per package.
Catalog No. 50-153-2366 Supplier Chemglass Life Sciences Supplier No. CLS4285LS1
Vented Cap Label for Reaction Tubes, 5 Labels per Row (1 thru 5), 50 Labels per Sheet. Labels are used to block from 1 to 5 openings on the stopper of the 50mL bio reaction tubes to control the amount of CO2 exchange and evaporation in the tube. Sheets have 8 each of white Mylar labels from numbers 1 to 5, and are 10 sets of labels per package.