Learn More
Air Science Usa Vented-Enclosure, 34" Width, Extra Tall Height, Polypropylene Construction (VE34T)
SDP
Catalog No. NC1837399
Encompass_Preferred
Vented Enclosures provide effective containment of airborne particulates during manipulation and transfer of potent compounds. Our in-house research suggests that even slight modifications of enclosures can result in containment factors from the microgram down to the nanogram. Our turbulent-free design utilizes environmentally friendly, ductless technology in combination with carbon / HEPA filtration to provide precise, safe containment in all applications. Ductless technology also makes installation of dedicated workstations easy, as no additional Hvac design is required. offers a host of vented enclosures to meet every analytical need. (VE34T)
Catalog No. NC1837399 Supplier Air Science Usa Supplier No. VE34T
Vented Enclosures provide effective containment of airborne particulates during manipulation and transfer of potent compounds. Our in-house research suggests that even slight modifications of enclosures can result in containment factors from the microgram down to the nanogram. Our turbulent-free design utilizes environmentally friendly, ductless technology in combination with carbon / HEPA filtration to provide precise, safe containment in all applications. Ductless technology also makes installation of dedicated workstations easy, as no additional Hvac design is required. offers a host of vented enclosures to meet every analytical need. (VE34T)