Learn More
CELLTREAT Scientific Products 150cm2 Tissue Culture Flask - Vent Cap, Sterile
SDP
Catalog No. NC3718272
Encompass_Preferred
Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachmentStable base enables filling with flask standing upright182cm2 flask offers more growth area than standard 175cm2 flasks300cm2 flask offers a large culture area with the convenience of a single layer flaskConvenient zip-closure bag is a practical alternative to folding and taping opened bagsVent caps feature 0.22μm hydrophobic membraneLarge opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapersMolded, interlocking T design for improved stacking - included on 75, 182 & 300cm² and 250, 600 & 850mL size flasks onlyFlask is printed with lot number and expiration date for easy identification and tracking150cm2 Flask features U-shaped design for more efficient cell harvesting"
Catalog No. NC3718272 Supplier CELLTREAT Scientific Products Supplier No. 229346
Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachmentStable base enables filling with flask standing upright182cm2 flask offers more growth area than standard 175cm2 flasks300cm2 flask offers a large culture area with the convenience of a single layer flaskConvenient zip-closure bag is a practical alternative to folding and taping opened bagsVent caps feature 0.22μm hydrophobic membraneLarge opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapersMolded, interlocking T design for improved stacking - included on 75, 182 & 300cm² and 250, 600 & 850mL size flasks onlyFlask is printed with lot number and expiration date for easy identification and tracking150cm2 Flask features U-shaped design for more efficient cell harvesting"