Learn More
Bolle Safety™ Vented Universal Safety Goggles
Catalog No. 17467444
Change view
Click to view available options
Style:
Clear
Clear and Foam
Clear, Foam, and Mouthguard
Smoke and Foam
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
17-467-444 Smoke and Foam
17-467-446 Clear and Foam
17-467-448 Clear, Foam, and Mouthguard
17-467-451 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-467-444 Supplier Bolle Safety™ Supplier No. UNIVGN30W
Only null left
Add to Cart
Add to Cart
This goggle provides optimum protection and comfort in all circumstances thanks to its ergonomic design. This innovative goggle with its exclusive platinum coating is available in several versions, each adapted to a specific work environment to offer more peace of mind and efficiency for the worker.
  • Optimal fit thanks to its Wave technology
  • PC/TPR co-injected frame with overflow chute
  • Effortless pivoting faceguard
  • Improved recyclability
  • UV protection
  • Scratch-resistant
  • Anti-fog
Specifications
Lens Coating Scratch-resistant, Anti-fog
Description Vented
Color Black and Yellow
Lens Tint Copper
Product Type Safety Goggles
Style Smoke and Foam
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
Frame Type Standard