Learn More
Research Products International Corp Safety Vented Solvent Wash Bottles, 16oz, Ethanol, 3 per Case
SDP
Catalog No. 50212585
Encompass
Wash Bottles feature patented venting assembly that prevents vapor pressure build-up of solvent within the wash bottle. Eliminates solvent drips from the spout due to internal evaporation. Wash Bottles are labelled with most commonly used solvent names. Bottles are low-density polyethylene, closures and venting assembly are polypropylene.
Catalog No. 50-212-585 Supplier Research Products International Corp Supplier No. 248672
Wash Bottles feature patented venting assembly that prevents vapor pressure build-up of solvent within the wash bottle. Eliminates solvent drips from the spout due to internal evaporation. Wash Bottles are labelled with most commonly used solvent names. Bottles are low-density polyethylene, closures and venting assembly are polypropylene.