Learn More
Brady™ Ventilated Lockable Confined Space Training Kits
Catalog No. 19129748 Shop All Brady Corporation Products
Change view
Click to view available options
Displayed Text:
Permit required. No unauthorized entry.
Diameter (English):
30 in.
Catalog No. Displayed Text Diameter (English)
19-129-748 Permit required. No unauthorized entry. 30 in.
1 options
Catalog No. 19-129-748 Supplier Brady™ Supplier No. 151058
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Visually identifying your confined spaces will keep you compliant, but it may leave gaps in safety. Add another level of safety with this Confined Space Safety Cover to keep your crew safe.
  • Get the complete safety package – unlike signs and tapes this cover provides a sign and an unavoidable boundary in one easy-to-use product
  • Ventilated covers are ideal for areas were noxious gasses are a concern, and include a 1.5 in. gas-testing port that allows for testing without removing the cover
  • Lock it down – get another level of safety with lockable options that give authorized personnel the option to apply a padlock to maintain restrictive access
  • Made of durable fabric for excellent resistance against oils and hydro-carbons
Specifications
Product Type Confined Space Cover
Color Black/Red
Material Metal
Diameter (English) 30 in.
Displayed Text Permit required. No unauthorized entry.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.