Learn More
Heidolph™ Ventilation and Replenishment Valve
SDP
Catalog No. 036210761 Shop All Heidolph North America Products
Change view
Click to view available options
Includes:
Valve
Catalog No. Includes
03-621-0761 Valve
1 options
Catalog No. 03-621-0761 Supplier Heidolph™ Supplier No. 036210761
Add to Cart
Add to Cart
For precise dosing during the process with venting function and greas-free ground joint.
For use with Hei-VAP Core, Hei-VAP Expert, Hei-VAP Ultimate, Hei-VAP Expert Control, and Hei-VAP Ultimate Control
Specifications
For Use With (Equipment) Hei-VAP Core, Hei-VAP Expert, Hei-VAP Ultimate, Hei-VAP Expert, Hei-VAP Expert Control, Hei-VAP Ultimate Control Rotary Evaporator
Includes Valve
Type Rotary Evaporator Accessory
Timer No
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov