Learn More
World Precision Instrument Ventilator for small animals
SDP
Catalog No. 502538873
Encompass
Advanced sensors for airflow and pressure insure the accuracy of respiratory parameters. The simple but versatile operating modes allow safe, long-term ventilation of small, delicate animals.
Remote computer control using the built-in USB port and a host PC is available. Basic control and monitoring software is provided, and specialized software packages are optional. Using such external control programs, it is possible to synchronize the ventilator for imaging purposes, do neural-controlled ventilation, constant-CO2 ventilation and much more.
Catalog No. 50-253-8873 Supplier World Precision Instrument Supplier No. CWSAR1000
Add to Cart
Add to Cart
Request a Quote
Advanced sensors for airflow and pressure insure the accuracy of respiratory parameters. The simple but versatile operating modes allow safe, long-term ventilation of small, delicate animals.
Remote computer control using the built-in USB port and a host PC is available. Basic control and monitoring software is provided, and specialized software packages are optional. Using such external control programs, it is possible to synchronize the ventilator for imaging purposes, do neural-controlled ventilation, constant-CO2 ventilation and much more.