Learn More
Andwin Scientific Filtered Venting Needle Rx-Vent Non-Coring Bevel 20 Gauge 1-1/2 Inch, 100/cs
SDP
Catalog No. 501692575
Encompass
Non-coring needle penetrates all stopper with minimum risk of damage. For venting evacuated contianers. Includes convenient sealed needle guard.
This item is not returnable. View return policy
Catalog No. 501692575 Supplier Andwin Scientific Supplier No. 3120
Non-coring needle penetrates all stopper with minimum risk of damage. For venting evacuated contianers. Includes convenient sealed needle guard.