Learn More
Fritsch Milling And Sizing Inc Venting Ring Wet Sieving 200 mm, V2A, Height 25 mm
SDP
Catalog No. 501534179
Encompass
Venting ring for test sieves 200 mm dia.required when utilizing test sieves < 100 µm. Venting rings are used to prevent the occurrence of air cushions during wet sieving.
Catalog No. 50-153-4179 Supplier Fritsch Milling And Sizing Inc Supplier No. 31.1330.03
Venting ring for test sieves 200 mm dia.required when utilizing test sieves < 100 µm. Venting rings are used to prevent the occurrence of air cushions during wet sieving.