Learn More
Pyramex™ Venture 3™ Safety Eyewear
Catalog No. 19155762 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Amber
Blue
Clear
Gray
IR/UV 5.0
Indoor/Outdoor, Mirror
Silver, Mirror
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-762 Clear
19-155-764 Gray
19-155-766 Amber
19-155-768 Blue
19-155-770 Silver, Mirror
19-155-771 Indoor/Outdoor, Mirror
19-155-767 IR/UV 5.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-155-762 missing translation for 'mfr' Pyramex™ missing translation for 'supplierNo' SB5710D
Add to Cart
Add to Cart
This item is not returnable. View return policy
Passes MIL-PRF 32432 High Velocity Impact Standards
  • Ventilated nose piece is soft and adjustable
  • Top of frame is ventilated allowing air to circulate
  • Soft dual-injected rubber temple tips combined with fully adjustable straight back temples reinforce the highest level of comfort and fit
  • 9.5 base curve lens provides excellent side protection
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Frame Color Black
    Weight (Metric) 28 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate