Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Venture Gear Earmuffs

Catalog No. 19555716 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Model:
VG9010
Color:
Light Blue
Packaging Type:
Clamshell
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Color Packaging Type
19-555-716 VG9010 Light Blue Clamshell
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-555-716 Supplier Pyramex™ Supplier No. VGPM9026IMP

Pyramex™ Venture Gear Earmuffs with ANSI S3.19 safety compliance

Venture Gear Earmuffs come in a variety of styles and colors to fit every need

  • BH9010 Series
    • Noise Reduction Rating (NRR) - 22 dB
    • Can be worn with hardhats, shields, and other PPE
    • Adjustable, durable headband
    • Large, soft foam earcups
    • Earcups tilt for optimum comfort and efficiency
  • VG10 Series
    • Noise Reduction Rating (NRR) - 25 dB
    • Dielectric
    • Large, soft foam ear cups
    • Adjustable, durable headband
  • VG80 Series
    • Noise Reduction Rating (NRR) - 36 dB
    • Low-profile ear cup design
    • Height can be adjusted to fit all sizes
    • Fold-away, padded headband
    • Packaged in a clamshell with free, disposable earplugs
  • VG90 Series
    • Noise Reduction Rating (NRR) - 22 dB
    • Adjustable, durable neckband
    • Large, soft foam earcups
    • Earcups tilt for optimum comfort and efficiency

Specifications

Color Light Blue
Certifications/Compliance ANSI S3.19
Size Universal
For Use With (Application) Hearing Protection
Model VG9010
Packaging Type Clamshell
Noise Reduction Rating (NRR) 22 dB
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.