Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Venture Gear Behind-the-Head Ear Muff

Catalog No. 19555708 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
22 dB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Noise Reduction Rating (NRR)
19-555-708 22 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-555-708 Supplier Pyramex™ Supplier No. VGBH9010C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venture Gear Behind-the-Head Ear Muffs are ideal for workers who wear or use additional PPE.

  • Can be worn with hard hats, face shields and other personal protective equipment
  • Adjustable and durable neckband
  • Large soft foam ear cups
  • Ear cups tilt for optimum comfort and efficiency

Specifications

Color Black, Green
ANSI S3.1974 Yes
Cushion Replacements No
For Use With (Application) Variety of industrial applications where additional PPE is worn or used.
High Visibility No
Model BH9010
Packaging Type Clamshell
Noise Reduction Rating (NRR) 22 dB
Corded No
Product Type Venture Gear
Disposable Reusable
Metal Detectable No
Headband Color Black
Dielectric No
Style Behind-the-head
Foldable No
EN 352.2002 EN 352-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.