Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Venture Gear VG10 Series Ear Muff

Catalog No. 19555709 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Model:
VG10
Color:
Black, Green
Packaging Type:
Clamshell
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Color Packaging Type
19-555-709 VG10 Black, Green Clamshell
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-555-709 Supplier Pyramex™ Supplier No. VGPM1010C

VG10 Series Ear Muffs are electrically insulated and feature a durable and adjustable headband.

  • Noise Reduction Rating (NRR) - 25 dB
  • Dielectric
  • Large, soft foam ear cups
  • Adjustable, durable headband

Specifications

Color Black, Green
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Variety of industrial applications
Model VG10
Packaging Type Clamshell
Noise Reduction Rating (NRR) 25 dB
Dielectric Yes
Style Over-the-head
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.