Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ AmpBT™ Series Electronic Bluetooth Ear Muff

Catalog No. 19555720 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-555-720 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-555-720 Supplier Pyramex™ Supplier No. VGPME30BT
Add to Cart
Add to Cart

AmpBT™ Electronic Ear Muffs compress harmful sounds to safe levels and is Bluetooth compatible for wireless listening and cell phone use.

  • Bluetooth compatible
  • Rubberized coating on ear cups
  • Automatic shut-off after four hours
  • 3.5 mm Aux jack for use of MP3 and mobile devices
  • Quad microphones
  • 2 AAA batteries included

Specifications

Color Black
ANSI S3.1974 Yes
Cushion Material Fabric, PVC
Cushion Color Black
Headband Material Polyurethane
For Use With (Application) Hearing Protection
Model VGPME30BT
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 26 dB
Headband Color Black
Dielectric No
Style Over-the-head
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.