Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Venture Gear Sentinel™ Electronic Ear Muffs

Catalog No. 19555719 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black
Light Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-555-719 Light Blue
19-555-718 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-555-719 Supplier Pyramex™ Supplier No. VGPME26
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sentinel™ Ear Muffs feature front-facing microphones and a 3.5 mm Aux jack for use with mobile devices.

  • Electronic ear muffs amplify hearing while suppressing harmful sounds.
  • Slim profile ear cups
  • Fold-away, padded headband
  • Front facing microphones
  • Automatic shut-off after four hours
  • 3.5 mm Aux jack for use of MP3 and mobile devices
  • 2 AAA Batteries Included

Specifications

Color Light Blue
ANSI S3.1974 Yes
Cushion Material PVC
Cushion Color Black
Headband Material Polyurethane
For Use With (Application) Hearing Protection
Model VGPME26
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 26 dB
Product Type Sentinel
Headband Color Black
Dielectric No
Style Over-the-head
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.