Learn More
Pyramex™ Venture II™ Readers Safety Eyewear
Catalog No. 19155663 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
Magnification Power:
+1.50
+2.00
+2.50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Magnification Power
19-155-663 Amber +2.50
19-155-647 Clear +1.50
19-155-649 Clear +2.00
19-155-651 Clear +2.50
19-155-661 Amber +1.50
19-155-662 Amber +2.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-155-663 Supplier Pyramex™ Supplier No. SB1830R25
Add to Cart
Add to Cart
This item is not returnable. View return policy
Temples adjust to four different lengths
  • Adjustable temples
  • Rubber nosepiece secures the glass and prevents slipping
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Amber
    Frame Color Black
    Magnification Power +2.50
    Weight (Metric) 27 g
    Lens Material Polycarbonate
    Frame Material Nylon 6.6