Learn More
Pyramex™ Venture II™ Safety Glasses
Catalog No. 19822693 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Blue
Metallic Blue
Lens Coating:
Anti-fog, Scratch-resistant, Impact-resistant, UV-treated
Scratch-resistant, Impact-resistant, UV-treated
Scratch-resistant, UV-treated, Impact-resistant
Lens Tint:
Amber
Clear
Gold Mirror
Gray
IR Filter Shade 3.0
IR Filter Shade 5.0
Indoor/Outdoor Mirror
Indoor/Outdoor Mirror, Indoor/Outdoor
Infinity Blue
Orange
Silver Mirror
Sun Block Bronze
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-822-693 Blue Scratch-resistant, Impact-resistant, UV-treated Gray
19-121-911 Black Scratch-resistant, Impact-resistant, UV-treated Orange
19-817-370 Black Scratch-resistant, Impact-resistant, UV-treated Gray
19-822-692 Metallic Blue Scratch-resistant, Impact-resistant, UV-treated Clear
19-151-450 Black Scratch-resistant, Impact-resistant, UV-treated Sun Block Bronze
19-151-448 Black Anti-fog, Scratch-resistant, Impact-resistant, UV-treated Gray
19-151-455 Black Scratch-resistant, Impact-resistant, UV-treated Gold Mirror
19-151-457 Black Scratch-resistant, UV-treated, Impact-resistant IR Filter Shade 3.0
19-151-456 Black Scratch-resistant, Impact-resistant, UV-treated IR Filter Shade 5.0
19-817-372 Black Scratch-resistant, UV-treated, Impact-resistant Silver Mirror
19-151-446 Black Anti-fog, Scratch-resistant, Impact-resistant, UV-treated Clear
19-051-451 Black Scratch-resistant, Impact-resistant, UV-treated Infinity Blue
19-817-374 Black Scratch-resistant, UV-treated, Impact-resistant Indoor/Outdoor Mirror
19-817-369 Black Scratch-resistant, UV-treated, Impact-resistant Clear
19-151-462 Metallic Blue Scratch-resistant, Impact-resistant, UV-treated Indoor/Outdoor Mirror, Indoor/Outdoor
19-822-691 Black Scratch-resistant, Impact-resistant, UV-treated Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-822-693 Supplier Pyramex™ Supplier No. SMB1820S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Venture II™ Safety Glasses feature full side protection without distracting sideshields or seams.
  • Lightweight spectacles integrate a sideshield into the lens
  • High-curve (9.75) dual-lens design provides more than 180° of coverage
  • Coated polycarbonate lenses provide superior scratch resistance and filter 99% of all harmful UV light
  • Temples adjust to four different sizes
  • Pillow-soft, thermoplastic elastomer ''nose-buds'' secure glasses on face for a comfortable fit
Compliance
Meet ANSI Z87.1 requirements and CSA Z94.3 standards
Specifications
Lens Coating Scratch-resistant, Impact-resistant, UV-treated
Certifications/Compliance Meet ANSI Z87.1 requirements and CSA Z94.3 standards
Lens Tint Gray
Product Type Safety Glasses
Frame Color Blue
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate, Nylon
UV Protection U6
Frame Type Half Frame
Autoclavable Autoclavable