Learn More
Pyramex™ VentureGear™ Drop Zone™ Safety Eyewear Kit
Catalog No. 19156056 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Impact-resistant
Catalog No. Lens Coating
19-156-056 Impact-resistant
1 options
This item is not returnable. View return policy
Catalog No. 19-156-056 Supplier Pyramex™ Supplier No. VGSB88KIT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Interchangeable lenses with black temples
  • Stylish contoured temples
  • Extremely lightweight
  • Rubber nosepiece for non-slip, all day comfort
  • Easy to change replacement lenses
  • Kit includes: Removable temples Crush-resistant, neoprene storage case
  • Four interchangeable H2X anti-fog lenses: Clear, Forest Gray, Orange, and Vermilion
  • Dropzone comes packaged in a free standing clamshell for countertop or pegboard display
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Lens Coating Impact-resistant
    Certifications/Compliance ANSI Z87.1,CAN/CSA Z94.3-07
    Certifications/Compliance (Europe) EN-166
    Includes Removable temples, Crush-resistant, neoprene storage case, Microfiber cleaning cloth, Four interchangeable lenses
    Lens Material Polycarbonate