Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Venture Gear Ever-Lite Range Combo Kit

Catalog No. 19156010 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black, Gray
Black, Pink
Model:
VGCOMBO8610
VGCOMBO8617
VGCOMBO8630
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Model
19-156-010 Black, Gray VGCOMBO8610
19-156-011 Black, Pink VGCOMBO8617
19-156-012 Black, Gray VGCOMBO8630
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-156-010 Supplier Pyramex™ Supplier No. VGCOMBO8610

PM80 Ear Muff with VentureGear™ Safety Glasses in a convenient combination package ideal for applications that require simultaneous hearing and eye protection.

  • Combination hearing/eyewear package
  • PM8010 Ear Muffs feature low-profile design, soft foam ear cups and fold-away padded headband
  • VentureGear™ Eyewear features black frame, soft nosepiece, wraparound lens, UVA/B/C protection
  • Available in three variations:
    • Gray ear muff/clear lens eyewear
    • Gray ear muff/amber lens eyewear
    • Pink ear muff/clear lens eyewear

Specifications

Color Black, Gray
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Variety of industrial and recreational applications
Model VGCOMBO8610
Pairs per Package 1 Pk.
Noise Reduction Rating (NRR) 26 dB
Product Type Ear Muff/Eyewear Combo Kit
Headband Color Black
Style Over-the-head
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.