Learn More
Pyramex™ VentureGear™ Howitzer™ Safety Eyewear
Catalog No. 19156080 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Tan
Lens Tint:
Bronze
Clear
Green
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-156-080 Tan Green
19-156-032 Black Clear
19-156-033 Black Bronze
19-156-034 Black Green
19-156-078 Tan Clear
19-156-079 Tan Bronze
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-156-080 Supplier Pyramex™ Supplier No. VGST1322T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Custom-fit rubber nosepiece and nonslip flexible co-injected temples
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Includes microfiber cleaning bag
  • Specifications
    Certifications/Compliance ANSI Z87.1,CSA Z94.3
    Lens Tint Green
    Frame Color Tan
    Weight (Metric) 35 g
    Lens Material Polycarbonate
    Frame Material Nylon