Learn More
Pyramex™ VentureGear™ Pagosa™ Safety Eyewear
Catalog No. 19156048 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
White
Lens Tint:
Blue, Mirror
Bronze
Green
Silver, Mirror
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-156-048 Black Green
19-156-049 Black Green
19-156-050 Black Blue, Mirror
19-156-047 Black Bronze
19-156-051 Black Silver, Mirror
19-156-086 White Bronze
19-156-087 White Green
19-156-088 White Blue, Mirror
19-156-089 White Blue, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-156-048 Supplier Pyramex™ Supplier No. VGSB522T
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Stylish, sunglass look safety glasses with wide, straightback temples
  • Wide straightback temples
  • Soft rubber nosepiece
  • Anti-fog, scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Green
    Frame Color Black
    Weight (Metric) 32 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate