Learn More
Pyramex™ VentureGear™ Semtex 2.0™ Safety Eyewear
Catalog No. 19156083 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Gun Metal
Tan
Lens Tint:
Bronze
Clear
Green
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-156-083 Tan Green
19-156-073 Gun Metal Clear
19-156-074 Gun Metal Bronze
19-156-075 Gun Metal Green
19-156-081 Tan Clear
19-156-082 Tan Bronze
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-156-083 Supplier Pyramex™ Supplier No. VGST1422T
Add to Cart
Add to Cart
This item is not returnable. View return policy
VentureGear™ Tactical
  • Custom-fit rubber nosepiece
  • Nonslip flexible co-injected temples
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Includes microfiber cleaning bag
  • Specifications
    Certifications/Compliance ANSI Z87.1,CSA Z94.3
    Lens Tint Green
    Frame Color Tan
    Weight (Metric) 31 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate