Learn More
Pyramex™ VentureGear™ Tensaw™ Safety Eyewear
Catalog No. 19156093 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
White, Blue
White, Gray
White, Red
Lens Tint:
Blue, Mirror
Red, Mirror
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-156-093 White, Blue Blue, Mirror
19-156-094 White, Red Red, Mirror
19-156-095 White, Red Red, Mirror
19-156-085 White, Gray Blue, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-156-093 Supplier Pyramex™ Supplier No. VGSWB365T
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Half-frame, high performance eyewear
  • Adjustable rubber nosepiece
  • Co-injected temples for snug, secure fit
  • Stylish half-frame high performance eyewear
  • Anti-fog, scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Blue, Mirror
    Frame Color White, Blue
    Weight (Metric) 30 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate