Learn More
Pyramex™ VentureGear™ Vallejo™ Safety Eyewear
Catalog No. 19156057 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Clear
Tortoise Shell
White
Lens Tint:
Blue, Mirror
Bronze
Green
Green, Mirror
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-156-057 Black Green
19-156-058 Black Green, Mirror
19-156-059 Black Blue, Mirror
19-156-067 Clear Blue, Mirror
19-156-084 Tortoise Shell Bronze
19-156-090 White Green
19-156-091 White Green, Mirror
19-156-092 White Blue, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-156-057 Supplier Pyramex™ Supplier No. VGSB922T
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Stylish full-frame high performance eyewear
  • Built-in rubber nosepiece
  • Stylish full-frame high performance eyewear
  • Vented frame increases air flow to prevent fogging
  • Anti-fog, scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Lens Coating Anti-fog
    Certifications/Compliance ANSI Z87.1
    Certifications/Compliance (Europe) EN-166
    Lens Tint Green
    Frame Color Black
    Weight (Metric) 31.7 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate, TPE