Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Venture Gear VG90 Series Ear Muffs

Catalog No. 19156020 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black, Gray
Black, Olive Drab Green
Black, Pink
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-156-020 Black, Gray
19-156-021 Black, Pink
19-555-714 Black, Olive Drab Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-156-020 Supplier Pyramex™ Supplier No. VGPM9010C
Add to Cart
Add to Cart

VG90 features a fold-away, padded headband and includes a pair of disposable ear plugs with each package.

  • Low-profile design
  • Soft foam ear cups
  • Height adjustment to fit all sizes
  • Fold-away, padded headband
  • Ear muffs are packaged with free disposable ear plugs
  • Ear muffs NRR 24 dB and increased to NRR 32 dB when used with ear plugs

Specifications

Color Black, Gray
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Variety of industrial and recreational applications
Model VG90
Packaging Type Clamshell
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 24 dB
Headband Color Black
Style Over-the-head
Foldable Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.