Learn More
Pyramex™ VentureGear™ Waverton Safety Eyewear
Catalog No. 19156060 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Brown
Lens Tint:
Blue, Mirror
Bronze
Green
Mirror
Silver, Mirror
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-156-060 Brown Bronze
19-156-027 Black Green
19-156-028 Black Blue, Mirror
19-156-029 Black Silver, Mirror
19-156-061 Brown Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-156-060 Supplier Pyramex™ Supplier No. VGSBR1118DB
Add to Cart
Add to Cart
This item is not returnable. View return policy
Packaged in a VentureGear™ branded polybag with retail tag
  • Flexible rubber nosepiece
  • Dual-injected straight-back temples
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Bronze
    Frame Color Brown
    Weight (Metric) 24.4 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate