Learn More
Enzo Life Sciences Venturicidin A (1mg). CAS: 33538-71-5
SDP
Catalog No. 502008975
Encompass_Preferred
Macrolide antibiotic. Potent inhibitor of mitochondrial ATP synthase complex, acting on the F0 membrane sector. Shows antifungal activity. Alternative name: Aabomycin A1. Formulation: Lyophilized. Purity: ≥95% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8975 Supplier Enzo Life Sciences Supplier No. ALX380211M001
Macrolide antibiotic. Potent inhibitor of mitochondrial ATP synthase complex, acting on the F0 membrane sector. Shows antifungal activity. Alternative name: Aabomycin A1. Formulation: Lyophilized. Purity: ≥95% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO. Long Term Storage: -20°C.