Learn More
AdipoGen Venturicidin A, 33538-71-5, MFCD00036323, 1 mg
SDP
Catalog No. 501146863
Encompass_Preferred
MF: C41H67NO11, MW: 750.0, ≥98%, Appearance: White solid. 20-Membered macrolide glycoside. Antibiotic and antifungal agent. Shows antitrypanosomal and antimalarial activity. Potent inhibitor of mitochondrial and bacterial ATP synthase. Inhibitor of F0F1-ATPase. Na+-translocating ATP synthases inhibitor.
Catalog No. 50-114-6863 Supplier AdipoGen Supplier No. BVT0454M001
MF: C41H67NO11, MW: 750.0, ≥98%, Appearance: White solid. 20-Membered macrolide glycoside. Antibiotic and antifungal agent. Shows antitrypanosomal and antimalarial activity. Potent inhibitor of mitochondrial and bacterial ATP synthase. Inhibitor of F0F1-ATPase. Na+-translocating ATP synthases inhibitor.