Learn More
Aviva Systems Biology Corp VENTX Antibody - middle region (ARP35877_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35877_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VENTX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human VENTX. Alternate names: HPX42B, MGC119910, MGC119911, NA88A, VENTX2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55904