Learn More
MSC Kwazar Handheld Sprayer, High Density Polyethylene, 1 L, Adjustable Nozzle, Viton Seals
Catalog No. 502838571
Encompass
These handheld sprayers are specifically designed for applications within the food industry and food processing. Constructed from high density polyethylene, they are made without dyes and are intended for safe contact with food, suitable for applying oils, fats, and food additives. Each sprayer has a 1 L capacity and features Viton seals along with an adjustable nozzle, providing versatility for various tasks.
  • Handheld sprayer type
  • High density polyethylene tank material
  • 1 L volume capacity
  • Mist and stream spray patterns
  • Chemical safe
  • Designed for food industry applications
  • In-tank filter
  • Viton seal material
  • Adjustable nozzle included
  • No hose
Catalog No. 50-283-8571 Supplier MSC Supplier No. 74333048
These handheld sprayers are specifically designed for applications within the food industry and food processing. Constructed from high density polyethylene, they are made without dyes and are intended for safe contact with food, suitable for applying oils, fats, and food additives. Each sprayer has a 1 L capacity and features Viton seals along with an adjustable nozzle, providing versatility for various tasks.
  • Handheld sprayer type
  • High density polyethylene tank material
  • 1 L volume capacity
  • Mist and stream spray patterns
  • Chemical safe
  • Designed for food industry applications
  • In-tank filter
  • Viton seal material
  • Adjustable nozzle included
  • No hose