Learn More
MSC Kwazar Compression Sprayer, Venus Super 360, 2.0 L, NBR Seals
Catalog No. 502835646
Encompass
The Venus Super 360 compression sprayer is designed for efficient and precise application of liquids. This 2.0 L capacity sprayer features a 360° system, allowing spraying from any tank position, making it suitable for various professional applications including cleaning, disinfection, and pest control.
  • System 360° enables spraying regardless of tank position.
  • Adjustable ending reaches hard-to-reach places.
  • Adjustable spray nozzle atomizes liquid from mist to stream.
  • Safety valve controls internal pressure.
  • Liquid level strip assists with easy dosing and monitoring.
  • Convenient handle provides ergonomic comfort.
  • Stabilizing ring protects the tank and adds stability.
Catalog No. 50-283-5646 Supplier MSC Supplier No. 74253386
The Venus Super 360 compression sprayer is designed for efficient and precise application of liquids. This 2.0 L capacity sprayer features a 360° system, allowing spraying from any tank position, making it suitable for various professional applications including cleaning, disinfection, and pest control.
  • System 360° enables spraying regardless of tank position.
  • Adjustable ending reaches hard-to-reach places.
  • Adjustable spray nozzle atomizes liquid from mist to stream.
  • Safety valve controls internal pressure.
  • Liquid level strip assists with easy dosing and monitoring.
  • Convenient handle provides ergonomic comfort.
  • Stabilizing ring protects the tank and adds stability.