Learn More
MSC Kwazar Sprayer, 1 L, High Density Polyethylene, Food Grade
Catalog No. 502836770
Encompass
This Kwazar Venus Pro+ Food Grade Sprayer is designed for use in the food industry and processing. It is made without dyes, intended for contact with food, and used for the application of oils, fats, and food additives. All seals are made of silicone.
  • Handheld sprayer
  • High Density Polyethylene tank material
  • 1 L volume capacity
  • Mist and stream spray pattern
  • Chemical safe
  • In-tank filter
  • Adjustable nozzle
  • No hose
  • Designed for use in the food industry and processing
  • Made without dyes
  • Intended for contact with food
  • Used in application of oils, fats, and food additives
  • All seals are made of silicone
Catalog No. 50-283-6770 Supplier MSC Supplier No. 74256009
This Kwazar Venus Pro+ Food Grade Sprayer is designed for use in the food industry and processing. It is made without dyes, intended for contact with food, and used for the application of oils, fats, and food additives. All seals are made of silicone.
  • Handheld sprayer
  • High Density Polyethylene tank material
  • 1 L volume capacity
  • Mist and stream spray pattern
  • Chemical safe
  • In-tank filter
  • Adjustable nozzle
  • No hose
  • Designed for use in the food industry and processing
  • Made without dyes
  • Intended for contact with food
  • Used in application of oils, fats, and food additives
  • All seals are made of silicone